accessibility menu, dialog, popup

UserName

CAS RN 1765-93-1

HOBOHF

CAS RN 1765-93-1

IUPAC Name: (4-fluorophenyl)boronic acid
Synonyms: 4-fluorophenylboronic acid
Molecular Weight (g/mol): 139.92
Molecular Formula: C6H6BFO2
InChi Key: LBUNNMJLXWQQBY-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)C=C1
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)