accessibility menu, dialog, popup

UserName

CAS RN 176442-21-0

IUPAC Name: 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-hydroxybenzoic acid
Synonyms: 3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-hydroxybenzoic acid
Molecular Weight (g/mol): 375.38
Molecular Formula: C22H17NO5
InChi Key: WGBZDECNYLZYRE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results