accessibility menu, dialog, popup

UserName

CAS RN 17610-21-8

H3COCH3H3CH3CCH3

CAS RN 17610-21-8

IUPAC Name: 1-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Synonyms: 1-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethanone
Molecular Weight (g/mol): 230.35
Molecular Formula: C16H22O
InChi Key: IHUSZOMIBSDQTB-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=C(C=C1)C(C)(C)CCC2(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results