accessibility menu, dialog, popup

UserName

CAS RN 17594-47-7

BaOOOOCCCH3CH3H3CH3CCH3H3CH3CCH3H3CCH3CH3H3CHH

CAS RN 17594-47-7

IUPAC Name: barium(2+) bis(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate) hydrate
Synonyms: barium(2+) bis(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate) hydrate
Molecular Weight (g/mol): 521.88
Molecular Formula: C22H40BaO5
InChi Key: VCALGUJWYYNHDY-UHFFFAOYSA-L
SMILES: O.[Ba++].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C
Boiling Point 
  • (4)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results