accessibility menu, dialog, popup

UserName

CAS RN 1759-28-0

CH3NSH2C

CAS RN 1759-28-0

IUPAC Name: 5-ethenyl-4-methyl-1,3-thiazole
Synonyms: thiazole, 5-ethenyl-4-methyl-
Molecular Weight (g/mol): 125.19
Molecular Formula: C6H7NS
InChi Key: QUAMMXIRDIIGDJ-UHFFFAOYSA-N
SMILES: CC1=C(SC=N1)C=C
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)