accessibility menu, dialog, popup

UserName

CAS RN 17556-18-2

OCl

CAS RN 17556-18-2

IUPAC Name: 6-chloro-1,2,3,4-tetrahydronaphthalen-2-one
Synonyms: 6-chloro-3,4-dihydro-1H-naphthalen-2-one
Molecular Weight (g/mol): 180.63
Molecular Formula: C10H9ClO
InChi Key: QQSYTUUAEZUAKL-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(CC(=O)CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results