accessibility menu, dialog, popup

UserName

CAS RN 17534-15-5

SHSH

CAS RN 17534-15-5

IUPAC Name: (2-sulfanidylphenyl)sulfanide
Synonyms: 1,2-benzenedithiol
Molecular Weight (g/mol): 140.22
Molecular Formula: C6H4S2
InChi Key: JRNVQLOKVMWBFR-UHFFFAOYSA-L
SMILES: [S-]C1=CC=CC=C1[S-]
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)