accessibility menu, dialog, popup

UserName

CAS RN 175334-72-2

ONSNH2

CAS RN 175334-72-2

IUPAC Name: 1,2-oxazole-5-carbothioamide
Synonyms: 1,2-oxazole-5-carbothioamide
Molecular Weight (g/mol): 128.15
Molecular Formula: C4H4N2OS
InChi Key: CEDRGDFENMZKCQ-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results