accessibility menu, dialog, popup

UserName

H3CBrI

CAS RN 175278-30-5

IUPAC Name: 4-bromo-2-ethyl-1-iodobenzene
Synonyms: 4-bromo-2-ethyl-1-iodobenzene
Molecular Weight (g/mol): 310.96
Molecular Formula: C8H8BrI
InChi Key: JWKQXPHYKRQLEJ-UHFFFAOYSA-N
SMILES: CCC1=C(I)C=CC(Br)=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results