accessibility menu, dialog, popup

UserName

CAS RN 175278-28-1

CH3FNH2O

CAS RN 175278-28-1

IUPAC Name: 5-fluoro-2-methylbenzamide
Synonyms: 5-fluoro-2-methylbenzamide
Molecular Weight (g/mol): 153.16
Molecular Formula: C8H8FNO
InChi Key: VPAFHFHYJHZKSR-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results