accessibility menu, dialog, popup

UserName

CAS RN 175278-19-0

CH3ONHNOFFF

CAS RN 175278-19-0

IUPAC Name: N-[4-cyano-2-(trifluoromethoxy)phenyl]acetamide
Synonyms: N-[4-cyano-2-(trifluoromethoxy)phenyl]acetamide
Molecular Weight (g/mol): 244.17
Molecular Formula: C10H7F3N2O2
InChi Key: RHBNAXXJVYFEEA-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C(OC(F)(F)F)C=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results