accessibility menu, dialog, popup

UserName

CAS RN 175278-00-9

OFFFI

CAS RN 175278-00-9

IUPAC Name: 1-iodo-2-(trifluoromethoxy)benzene
Synonyms: 1-iodo-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 288.01
Molecular Formula: C7H4F3IO
InChi Key: GYBMJVZOZTVDKS-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results