accessibility menu, dialog, popup

UserName

CAS RN 175277-76-6

IUPAC Name: (2,4-difluorophenyl)thiourea
Synonyms: 2,4-difluorophenylthiourea
Molecular Weight (g/mol): 188.2
Molecular Formula: C7H6F2N2S
InChi Key: DZZSKQFBAGZNSH-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results