accessibility menu, dialog, popup

UserName

CH3OONNClFFF

CAS RN 175277-73-3

IUPAC Name: ethyl 6-chloro-5-cyano-2-(trifluoromethyl)pyridine-3-carboxylate
Synonyms: ethyl 6-chloro-5-cyano-2-(trifluoromethyl)pyridine-3-carboxylate
Molecular Weight (g/mol): 278.62
Molecular Formula: C10H6ClF3N2O2
InChi Key: PMDLSQHFIADYJD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N=C(Cl)C(=C1)C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results