accessibility menu, dialog, popup

UserName

CAS RN 175277-60-8

NOFFFN

CAS RN 175277-60-8

IUPAC Name: 3-[2-oxo-3-(trifluoromethyl)-1,2-dihydropyridin-1-yl]propanenitrile
Synonyms: 3-[2-oxo-3-(trifluoromethyl)pyridin-1-yl]propanenitrile
Molecular Weight (g/mol): 216.16
Molecular Formula: C9H7F3N2O
InChi Key: CQBBWSPMEJVWQE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CN(CCC#N)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results