accessibility menu, dialog, popup

UserName

CAS RN 175277-52-8

NClClFFF

CAS RN 175277-52-8

IUPAC Name: 3-chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine
Synonyms: 3-chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 230.01
Molecular Formula: C7H4Cl2F3N
InChi Key: NUSXEDNPFNWFDQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(CCl)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results