accessibility menu, dialog, popup

UserName

CAS RN 175276-79-6

NSNH2

CAS RN 175276-79-6

IUPAC Name: 3-(1H-pyrrol-1-yl)benzene-1-carbothioamide
Synonyms: 3-(pyrrol-1-yl)benzenecarbothioamide
Molecular Weight (g/mol): 202.28
Molecular Formula: C11H10N2S
InChi Key: LKZUYCBQOPNMNQ-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(=CC=C1)N1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results