accessibility menu, dialog, popup

UserName

CH3NH3CH3COHO

CAS RN 175276-50-3

IUPAC Name: 1,2,5-trimethyl-1H-pyrrole-3-carboxylic acid
Synonyms: 1,2,5-trimethylpyrrole-3-carboxylic acid
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: JTEBLTWGSAXWEE-UHFFFAOYSA-N
SMILES: CN1C(C)=CC(C(O)=O)=C1C
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results