accessibility menu, dialog, popup

UserName

CAS RN 175205-85-3

H2NOBrBr

CAS RN 175205-85-3

IUPAC Name: 3,5-dibromobenzamide
Synonyms: 3,5-dibromobenzamide
Molecular Weight (g/mol): 278.93
Molecular Formula: C7H5Br2NO
InChi Key: IOFGMNBDIDEBIQ-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(Br)=CC(Br)=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results