accessibility menu, dialog, popup

UserName

CAS RN 175205-56-8

FFFClICl

CAS RN 175205-56-8

IUPAC Name: 1,3-dichloro-2-iodo-5-(trifluoromethyl)benzene
Synonyms: 1,3-dichloro-2-iodo-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 340.89
Molecular Formula: C7H2Cl2F3I
InChi Key: RLCCFAVBZGICHD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(I)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results