accessibility menu, dialog, popup

UserName

ClClNNNN

CAS RN 175205-12-6

IUPAC Name: 5-(2,3-dichlorophenyl)-2H-1,2,3,4-tetrazol-2-ide
Synonyms: 5-(2,3-dichlorophenyl)-2H-1,2,3,4-tetrazol-2-ide
Molecular Weight (g/mol): 214.03
Molecular Formula: C7H3Cl2N4
InChi Key: IQJBSNPEVBTDMM-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(C2=N[N-]N=N2)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results