accessibility menu, dialog, popup

UserName

CAS RN 175137-04-9

CH3H3CCH3SOH2NNH2

CAS RN 175137-04-9

IUPAC Name: 3-amino-5-tert-butylthiophene-2-carboxamide
Synonyms: 3-amino-5-tert-butylthiophene-2-carboxamide
Molecular Weight (g/mol): 198.28
Molecular Formula: C9H14N2OS
InChi Key: MKHZJBZWKNDDCN-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(N)=C(S1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results