accessibility menu, dialog, popup

UserName

CAS RN 175137-03-8

CH3H3CCH3SOOH3CNH2

CAS RN 175137-03-8

IUPAC Name: methyl 3-amino-5-tert-butylthiophene-2-carboxylate
Synonyms: methyl 3-amino-5-tert-butylthiophene-2-carboxylate
Molecular Weight (g/mol): 213.3
Molecular Formula: C10H15NO2S
InChi Key: WHJBGOQJMFAKHY-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=C(S1)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results