accessibility menu, dialog, popup

UserName

CAS RN 175136-66-0

FFFFFFOFFFFFF

CAS RN 175136-66-0

IUPAC Name: bis[3,5-bis(trifluoromethyl)phenyl]methanone
Synonyms: bis[3,5-bis(trifluoromethyl)phenyl]methanone
Molecular Weight (g/mol): 454.22
Molecular Formula: C17H6F12O
InChi Key: GATWMPGNBWPCIY-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)C(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results