accessibility menu, dialog, popup

UserName

CAS RN 175135-67-8

ONOSN

CAS RN 175135-67-8

IUPAC Name: 2-(benzylsulfanyl)-5-nitrobenzonitrile
Synonyms: 2-(benzylsulfanyl)-5-nitrobenzonitrile
Molecular Weight (g/mol): 270.31
Molecular Formula: C14H10N2O2S
InChi Key: UVENNUQTKADSQA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(SCC2=CC=CC=C2)C(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results