accessibility menu, dialog, popup

UserName

CAS RN 175135-27-0

CH3SNONH2

CAS RN 175135-27-0

IUPAC Name: 2-(ethylsulfanyl)pyridine-3-carboxamide
Synonyms: 2-(ethylsulfanyl)pyridine-3-carboxamide
Molecular Weight (g/mol): 182.24
Molecular Formula: C8H10N2OS
InChi Key: YSSWIQWINOZESS-UHFFFAOYSA-N
SMILES: CCSC1=C(C=CC=N1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results