accessibility menu, dialog, popup

UserName

H2NNHOS

CAS RN 175135-07-6

IUPAC Name: 1-benzothiophene-2-carbohydrazide
Synonyms: 1-benzothiophene-2-carbohydrazide
Molecular Weight (g/mol): 192.24
Molecular Formula: C9H8N2OS
InChi Key: ZXKPFIRPUUAAPQ-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC2=CC=CC=C2S1
Percent Purity 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)