accessibility menu, dialog, popup

UserName

CAS RN 175135-06-5

IUPAC Name: 3,5-dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride
Synonyms: 3,5-dichloro-4-(2-chloro-4-nitrophenoxy)benzenesulfonyl chloride
Molecular Weight (g/mol): 417.03
Molecular Formula: C12H5Cl4NO5S
InChi Key: YNPAAILIMIGQIH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(OC2=C(Cl)C=C(C=C2Cl)S(Cl)(=O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results