CAS RN 17475-68-2
CAS RN 17475-68-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hafnium 1,1,1-trifluoro-2,4-pentanedionate, Thermo Scientific™
CAS: 17475-68-2 Molecular Formula: C20H16F12HfO8 Molecular Weight (g/mol): 790.81 MDL Number: MFCD00078027 InChI Key: JTUDTOWCQOQZBH-UHFFFAOYSA-N Synonym: Hafnium trifluoroacetylacetonate IUPAC Name: hafnium(4+) tetrakis(1,1,1-trifluoro-2,4-dioxopentan-3-ide) SMILES: [Hf+4].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F