accessibility menu, dialog, popup

UserName

OOOOOO

CAS RN 17455-13-9

IUPAC Name: 1,4,7,10,13,16-hexaoxacyclooctadecane
Synonyms: 18-crown-6 ether cyclo peg-6 ethylene oxide cyclic hexamer
Molecular Weight (g/mol): 264.32
Molecular Formula: C12H24O6
InChi Key: XEZNGIUYQVAUSS-UHFFFAOYSA-N
SMILES: C1COCCOCCOCCOCCOCCO1
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (7)
Physical Form 
  • (4)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (7)
  • (4)
  • (3)
  • (2)