accessibility menu, dialog, popup

UserName

FFFNCS

CAS RN 1743-86-8

IUPAC Name: 1-isothiocyanato-2-(trifluoromethyl)benzene
Synonyms: 1-isothiocyanato-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 203.18
Molecular Formula: C8H4F3NS
InChi Key: FCEKLQPJGXIQRY-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1N=C=S
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results