accessibility menu, dialog, popup

UserName

CAS RN 17422-32-1

ClHN

CAS RN 17422-32-1

IUPAC Name: 5-chloro-1H-indole
Synonyms: 5-chloro-1H-indole
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: MYTGFBZJLDLWQG-UHFFFAOYSA-N
SMILES: ClC1=CC=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)