accessibility menu, dialog, popup

UserName

CH3ONOO

CAS RN 17408-16-1

IUPAC Name: 1-(3-nitrophenyl)propan-1-one
Synonyms: 1-(3-nitrophenyl)propan-1-one
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: VSPOTMOYDHRALZ-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results