accessibility menu, dialog, popup

UserName

CAS RN 17372-87-1

IUPAC Name: disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Synonyms: eosin Y eosin YS dye eosine yellowish acid red 87 eosine G
Molecular Weight (g/mol): 691.86
Molecular Formula: C20H6Br4Na2O5
InChi Key: SEACYXSIPDVVMV-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (19)
Quantity 
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
Color 
  • (1)
  • (3)
  • (5)
  • (3)
Grade 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (7)
  • (11)
  • (3)
  • (1)
  • (1)
  • (19)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (7)
  • (2)
  • (2)
  • (2)