accessibility menu, dialog, popup

UserName

CAS RN 1737-36-6

IUPAC Name: 4-chloro-3-(trifluoromethyl)benzoate
Synonyms: 4-chloro-3-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 223.56
Molecular Formula: C8H3ClF3O2
InChi Key: PPHHAZOVVZBSCM-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(Cl)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results