accessibility menu, dialog, popup

UserName

CH3OONONO

CAS RN 17348-71-9

IUPAC Name: 5-(ethoxycarbonyl)-2,1,3-benzoxadiazol-1-ium-1-olate
Synonyms: 5-(ethoxycarbonyl)-2,1,3-benzoxadiazol-1-ium-1-olate
Molecular Weight (g/mol): 208.17
Molecular Formula: C9H8N2O4
InChi Key: PNSWPHLJTBSRJR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=NO[N+]([O-])=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results