CAS RN 17345-77-6
IUPAC Name:
4-bromobenzene-1,2-diol
Synonyms:
4-bromocatechol C6H5BrO2
Molecular Weight (g/mol):
189.01
Molecular Formula:
C6H5BrO2
InChi Key:
AQVKHRQMAUJBBP-UHFFFAOYSA-N
SMILES:
OC1=CC=C(Br)C=C1O
1
–
1
of
1
results
1
4-Bromocatechol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.