accessibility menu, dialog, popup

UserName

CAS RN 17316-55-1

H3COPOOCH3(E/Z)

CAS RN 17316-55-1

IUPAC Name: diethyl (3-phenylprop-2-en-1-yl)phosphonate
Synonyms: diethyl 3-phenylprop-2-en-1-ylphosphonate
Molecular Weight (g/mol): 254.27
Molecular Formula: C13H19O3P
InChi Key: AWKOMEXJXBINBQ-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC=CC1=CC=CC=C1)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results