CAS RN 17311-31-8
CAS RN 17311-31-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dioxidine, TRC
CAS: 17311-31-8 Molecular Formula: C10 H10 N2 O4 Molecular Weight (g/mol): 222.197 Synonym: 2,3-Bis(hydroxymethyl)quinoxaline-1,4-di-N-oxide,2,3-Di(hydroxymethyl)quinoxaline 1,4-Di-N-oxide,2,3-Di(hydroxymethyl)quinoxaline Di-N-oxide,Dioxydine,Farmoxidin,Mastoxidine,Metrosept; IUPAC Name: [3-(hydroxymethyl)-1,4-dioxo-quinoxalin-2-yl]methanol SMILES: OCc1c(CO)n(=O)c2ccccc2n1=O