accessibility menu, dialog, popup

UserName

H3CCH3H3COONHO

CAS RN 172647-87-9

IUPAC Name: tert-butyl 2-(hydroxymethyl)-2,3-dihydro-1H-indole-1-carboxylate
Synonyms: tert-butyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate
Molecular Weight (g/mol): 249.31
Molecular Formula: C14H19NO3
InChi Key: JNQNQSCDMKNYEX-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1C(CO)CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results