accessibility menu, dialog, popup

UserName

CAS RN 17243-13-9

SOOClOHOOH

CAS RN 17243-13-9

IUPAC Name: 5-(chlorosulfonyl)-2-hydroxybenzoic acid
Synonyms: 5-(chlorosulfonyl)-2-hydroxybenzoic acid
Molecular Weight (g/mol): 236.62
Molecular Formula: C7H5ClO5S
InChi Key: SJXHSFSHNKFRLN-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(O)C=CC(=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results