accessibility menu, dialog, popup

UserName

CAS RN 172324-67-3

OO

CAS RN 172324-67-3

IUPAC Name: 3-[(benzyloxy)methyl]cyclobutan-1-one
Synonyms: 3-[(benzyloxy)methyl]cyclobutan-1-one
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: FHBBBGYOVFMVIB-UHFFFAOYSA-N
SMILES: O=C1CC(COCC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results