accessibility menu, dialog, popup

UserName

H3COOCH3ClO

CAS RN 17213-57-9

IUPAC Name: 3,5-dimethoxybenzoyl chloride
Synonyms: 3,5-dimethoxybenzoyl chloride
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: FTHPLWDYWAKYCY-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)