CAS RN 17200-29-2
CAS RN 17200-29-2
IUPAC Name:
5-chloro-1,3-benzoxazole
Synonyms:
benzoxazole, 5-chloro-
Molecular Weight (g/mol):
153.57
Molecular Formula:
C7H4ClNO
InChi Key:
VWMQXAYLHOSRKA-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2OC=NC2=C1