accessibility menu, dialog, popup

UserName

CAS RN 1720-38-3

IUPAC Name: deca-1,9-diyne
Synonyms: 1,9-decadiyne
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: ILVDYAGPHFWNQI-UHFFFAOYSA-N
SMILES: C#CCCCCCCC#C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)