CAS RN 171628-01-6
CAS RN 171628-01-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-[[[[[[[(Acetylamino)carbonyl]amino]carbonyl]amino]carbonyl]amino]sulfonyl]-2-thiophenecarboxylic Acid Methyl Ester (Technical Grade), TRC
CAS: 171628-01-6 Molecular Formula: C11H12N4O8S2 Molecular Weight (g/mol): 392.36 Synonym: Methyl 3-(N-(((acetylcarbamoyl)carbamoyl)carbamoyl)sulfamoyl)thiophene-2-carboxylate,Impurity of Thiophanate-methyl IUPAC Name: methyl 3-(acetylcarbamoylcarbamoylcarbamoylsulfamoyl)thiophene-2-carboxylate SMILES: COC(=O)c1sccc1S(=O)(=O)NC(=O)NC(=O)NC(=O)NC(=O)C