accessibility menu, dialog, popup

UserName

CAS RN 17147-85-2

CH3NOH3COO

CAS RN 17147-85-2

IUPAC Name: 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate
Synonyms: 3-ethyl-5-methyl-1,2-oxazole-4-carboxylate
Molecular Weight (g/mol): 154.15
Molecular Formula: C7H8NO3
InChi Key: RSWFHHWVFZWIMV-UHFFFAOYSA-M
SMILES: CCC1=NOC(C)=C1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results