accessibility menu, dialog, popup

UserName

CAS RN 171364-85-5

BOOH3CCH3H3CH3CN

CAS RN 171364-85-5

IUPAC Name: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Weight (g/mol): 255.12
Molecular Formula: C15H18BNO2
InChi Key: ARRJAONCYUAPJR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C2C=CC=CC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results