accessibility menu, dialog, popup

UserName

NSOON

CAS RN 170449-34-0

IUPAC Name: 2-(pyridine-2-sulfonyl)acetonitrile
Synonyms: 2-(pyridine-2-sulfonyl)acetonitrile
Molecular Weight (g/mol): 182.2
Molecular Formula: C7H6N2O2S
InChi Key: SAKACXZDAJXBCI-UHFFFAOYSA-N
SMILES: O=S(=O)(CC#N)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results