CAS RN 1703747-47-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gemfibrozil 1-O-Beta-Glucuronide-d6, TRC
CAS: 1703747-47-0 Molecular Formula: C21H24D6O9 Molecular Weight (g/mol): 432.49 Synonym: 1-[5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoate] β-D-Glucopyranuroic Acid-d6,Gemfibrozil-d6 Glucuronide; SMILES: CC1=C(OCCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C(O[C@@H]2O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]2O)=O)C=C(C)C=C1